2-(2-Hydroxyethyl)-1-methylpyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2-pyrrolidineethanol

A126283-1G A126283-5G A126283-10G A126283-100G A126283-5G A126283-25G A126283-1G
103800-788EA 9 USD
103800-788 77312-990 103831-642 76810-932 76810-930 77312-992 77312-994
2-(2-Hydroxyethyl)-1-methylpyrrolidine 98%
2-(2-Hydroxyethyl)-1-methylpyrrolidine

Caution: for Research Use Only

Formula: C₇H₁₅NO
MW: 129.20 g/mol
Boiling Pt: 214 °C (760 mm Hg)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003174
CAS Number: 67004-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR