4-Iodo-3-nitroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Iodo-4-methoxy-2-nitrobenzene

A194832-25G A194832-5G A194832-100G A194832-10G
103847-308EA 9 USD
103847-308 77085-236 77085-238 77139-212 77139-214
4-Iodo-3-nitroanisole 97%
4-Iodo-3-nitroanisole

Caution: for Research Use Only

Formula: C₇H₆INO₃
MW: 279.03 g/mol
Boiling Pt: 330.8 °C (760 mm Hg)
Melting Pt: 61…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00024195
CAS Number: 58755-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR