3-Amino-1H-pyrazole 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole
A331641-25G
A331641-10G
A331641-5G
103836-548EA
6
USD
103836-548
76839-540
77290-464
77306-414
3-Amino-1H-pyrazole 98%
3-Amino-1H-pyrazole
Caution: for Research Use Only
Formula:
C₃H₅N₃ Boiling Pt: 288.4 °C (760 mm Hg) Melting Pt: 34…37 °C Flash Pt: 182.569 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005236 CAS Number: 1820-80-0 UN: 3263 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...