1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid

A218770-10G A218770-250MG A218770-1G A218770-25G A218770-5G
103818-746EA 9 USD
103818-746 103849-616 76873-864 76873-866 77088-144 77088-146 77088-148
1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%
1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Caution: for Research Use Only

Formula: C₁₃H₉F₂NO₃
Boiling Pt: 434.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01646375
CAS Number: 93107-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR