2-Bromo-5-methoxybenzyl bromide 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Bromo-3-(bromomethyl)anisole, 1-Bromo-2-(bromomethyl)-4-methoxybenzene
A512317-1G
A512317-25G
A512317-5G
A512317-10G
103828-410EA
9
USD
103828-410
103839-510
76843-496
76843-498
76843-500
77034-612
2-Bromo-5-methoxybenzyl bromide 97%
2-Bromo-5-methoxybenzyl bromide
Caution: for Research Use Only
Formula:
C₈H₈Br₂O MW: 279.96 g/mol Boiling Pt: 299.6 °C (760 mm Hg) Melting Pt: 90…94 °C Flash Pt: 120.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078664 CAS Number: 19614-12-1 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...