4-Bromo-1-naphthylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-4-bromonaphthalene

A184009-25G A184009-5G A184009-100G A184009-10G A184009-1G
103815-698EA 6 USD
103815-698 103846-258 77083-872 77083-874 77191-778 77191-780 77301-874
4-Bromo-1-naphthylamine 98%
4-Bromo-1-naphthylamine

Caution: for Research Use Only

Formula: C₁₀H₈BrN
Boiling Pt: 338.4 °C (760 mm Hg)
Melting Pt: 102…103 °C
Flash Pt: 158.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004023
CAS Number: 2298-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR