1-Acetyladamantane 98%
Supplier: Ambeed
|
Warning
|
A151135-100G
A151135-10G
A151135-1G
A151135-500G
A151135-25G
A151135-5G
103842-382EA
9
USD
103842-382
77190-794
77190-796
77190-798
77190-800
77299-852
77299-854
1-Acetyladamantane 98%
1-Acetyladamantane
Caution: Sealed in dry, at room temperature.
Formula:
C₁₂H₁₈O Boiling Pt: 262.3 °C (760 mm Hg) Melting Pt: 53…55 °C Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074739 CAS Number: 1660-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...