10-Aminodecanoic acid 97%

Supplier: Ambeed
Warning

A287245-1G A287245-5G A287245-100MG A287245-250MG
103813-740EA 34 USD
103813-740 103834-880 76837-420 76837-422 77194-240 77194-242
10-Aminodecanoic acid 97%
10-Aminodecanoic acid

Caution: for Research Use Only

Formula: C₁₀H₂₁NO₂
Boiling Pt: 319.8 °C (760 mm Hg)
Flash Pt: 147.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00270349
CAS Number: 13108-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR