1,4-Dimethoxybenzene 98%
Supplier: Ambeed
|
Warning
|
A754084-100G
A754084-25G
A754084-500G
A754084-1000G
103810-554EA
58
USD
103810-554
76807-170
76807-172
76807-174
77308-258
1,4-Dimethoxybenzene 98%
1,4-Dimethoxybenzene
Caution: for Research Use Only
Formula:
C₈H₁₀O₂ Boiling Pt: 213 °C (760 mm Hg) Melting Pt: 54…56 °C Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008401 CAS Number: 150-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...