1,2-O-Isopropylidene-ɑ-D-xylofuranose 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-O-Isopropylidene-α-D-xylofuranose, (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol, (3aR,5R,6S,6aR)-5-(Hydroxymethyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

A167664-1G A167664-25G A167664-5G A167664-100G A167664-10G
103806-208EA 5 USD
103806-208 103844-466 77081-492 77081-494 77081-496 77191-320 77191-322
1,2-O-Isopropylidene-ɑ-D-xylofuranose 98%
1,2-O-Isopropylidene-alpha-D-xylofuranose

Caution: for Research Use Only

Formula: C₈H₁₄O₅
Boiling Pt: 333.0±37.0 °C (760 mm Hg)
Melting Pt: 69…71 °C
Flash Pt: 155.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00063295
CAS Number: 20031-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR