5-Amino-1,2,3-thiadiazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2,3-Thiadiazol-5-amine, 5-Amino-1,2,3-Thiadiazole
A242670-100G
A242670-250MG
A242670-1G
A242670-5G
103811-482EA
8
USD
103811-482
103851-300
77000-470
77000-472
77304-248
77304-250
5-Amino-1,2,3-thiadiazole 98%
5-Amino-1,2,3-thiadiazole
Caution: for Research Use Only
Formula:
C₂H₃N₃S Melting Pt: 224…235 °C Flash Pt: 98.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082810 CAS Number: 4100-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...