1,10-Phenanthroline-5,6-dione 98%

Supplier: Ambeed
Warning

A204705-1G A204705-25G A204705-5G A204705-10G A204705-250MG
103817-564EA 12 USD
103817-564 103848-228 77086-404 77086-406 77086-408 77192-416 77192-418
1,10-Phenanthroline-5,6-dione 98%
1,10-Phenanthroline-5,6-dione

Caution: for Research Use Only

Formula: C₁₂H₆N₂O₂
Boiling Pt: 456.1 °C (760 mm Hg)
Melting Pt: 260 °C
Flash Pt: 229.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00014473
CAS Number: 27318-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR