1,10-Phenanthroline 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
o-Phenanthroline
A326753-10G
A326753-25G
A326753-5G
A326753-100G
103836-396EA
8
USD
103836-396
76724-064
76839-362
76839-364
77601-470
1,10-Phenanthroline 98%
1,10-Phenanthroline
Caution: for Research Use Only
Formula:
C₁₂H₈N₂ MW: 180.21 g/mol Boiling Pt: 360 Melting Pt: 97…102 °C Flash Pt: 330 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011678 CAS Number: 66-71-7 UN: 2811 ADR: 6.1 Ⅲ Merck Index: 12,07356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...