1,1,7,7-Tetramethyl-8-hydroxyjulolidine 98%

Supplier: Ambeed
Warning

A167338-250MG A167338-1G
103806-168EA 8 USD
103806-168 103844-428 76690-082 77081-438
1,1,7,7-Tetramethyl-8-hydroxyjulolidine 98%
1,1,7,7-Tetramethyl-8-hydroxyjulolidine

Caution: for Research Use Only

Formula: C₁₆H₂₃NO
Storage Temperature: Ambient
MDL Number: MFCD09264711
CAS Number: 115704-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR