1-(Aminomethyl)cyclobutanol 97%

Supplier: Ambeed
Warning

Synonyms: 1-(aminomethyl)cyclobutan-1-ol

A343288-5G A343288-250MG A343288-25G
103825-880EA 59 USD
103825-880 103836-854 76839-904 77033-046 77033-048 77872-510
1-(Aminomethyl)cyclobutanol 97%
1-(Aminomethyl)cyclobutanol

Caution: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.14 g/mol
Boiling Pt: 176 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11040537
CAS Number: 180205-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR