1-(4-Aminophenyl)-4-(4-hydroxyphenyl)piperazine 98%

Supplier: Ambeed
Warning

A479529-1G A479529-25G A479529-5G A479529-10G
103828-114EA 5 USD
103828-114 103839-188 76843-126 76843-128 76843-130 77034-338
1-(4-Aminophenyl)-4-(4-hydroxyphenyl)piperazine 98%
1-(4-Aminophenyl)-4-(4-hydroxyphenyl)piperazine

Caution: for Research Use Only

Formula: C₁₆H₁₉N₃O
Boiling Pt: 532.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02093082
CAS Number: 74853-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR