(S)-1-Boc-3-hydroxypiperidine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected

A118676-100G A118676-10G A118676-1G A118676-25G A118676-500G A118676-5G
103830-464EA 9 USD
103830-464 76766-320 76766-322 76766-324 76766-326 76766-328 77311-766
(S)-1-Boc-3-hydroxypiperidine 98%
(S)-1-Boc-3-hydroxypiperidine

Caution: for Research Use Only

Formula: C₁₀H₁₉NO₃
Melting Pt: 34…40 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115307
CAS Number: 143900-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR