L(+)-Ascorbic acid 98%

Supplier: Ambeed
Warning

Synonyms: Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

A182821-1000G A182821-100G A182821-500G A182821-25G
103846-132EA 5 USD
103846-132 76974-510 76974-512 76974-514 77301-802
L(+)-Ascorbic acid 98%
L(+)-Ascorbic acid

Caution: for Research Use Only

Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553
Melting Pt: 190…194 °C
Flash Pt: 238.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
Merck Index: 13,00837

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR