Lycopene 98%

Supplier: Ambeed
Warning

Synonyms: (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-Octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene

A306463-1G A306463-50MG A306463-250MG A306463-1MG
103814-466EA 29 USD
103814-466 76838-342 77114-538 77114-540 77114-542 77612-758 77685-873
Lycopene 98%
Lycopene

Caution: for Research Use Only

Formula: C₄₀H₅₆
MW: 536.88 g/mol
Melting Pt: 172…173 °C
Storage Temperature: Freezer
MDL Number: MFCD00017350
CAS Number: 502-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR