4-Iodophenylboronic acid 97%

Supplier: Ambeed
Warning

A159270-250MG A159270-10G
103805-238EA 8 USD
103805-238 103843-456 77273-236 77550-002
4-Iodophenylboronic acid 97%
4-Iodophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₆BIO₂
Boiling Pt: 333.1 °C (760 mm Hg)
Melting Pt: 326…330 °C
Flash Pt: 155.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01319014
CAS Number: 5122-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR