4-Bromophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromobenzeneboronic acid, (4-Bromophenyl)boronic acid, Dihydroxy(4-bromophenyl)borane, (4-Bromophenyl)metaboric acid

A171431-25G A171431-5G A171431-100G A171431-10G A171431-500G
103806-634EA 5 USD
103806-634 103844-908 77082-066 77082-068 77111-030 77111-032 77111-034
4-Bromophenylboronic acid 97%
4-Bromophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₆BBrO₂
Boiling Pt: 315 °C (760 mm Hg)
Melting Pt: 280 °C
Flash Pt: 144.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00002104
CAS Number: 5467-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR