L(-)-Glutathione (oxidised form) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)
A397523-1G
A397523-25G
A397523-250MG
A397523-100G
103826-998EA
15
USD
103826-998
103838-038
76841-464
76841-466
76979-022
77672-962
L(-)-Glutathione (oxidised form) 95%
L(-)-Glutathione (oxidized form)
Caution: For research use only
Formula:
C₂₀H₃₂N₆O₁₂S₂ Boiling Pt: 1196.5 °C (760 mm Hg) Melting Pt: 178…182 °C Flash Pt: 677.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063106 CAS Number: 27025-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...