Cole-Parmer® CB-200 Series Block Chiller/Heater, 100 - 230 VAC
Supplier: Antylia Scientific
36620-17
490010-886EA 1848.04 USD
490010-886
Cole-Parmer® CB-200 Series Block Chiller/Heater, 100 - 230 VAC
Block Heaters Block Heater/Coolers
Safely maintain biological samples.
  • Designed for maintaining biological samples safely between 4 and 37 °C
  • Alarm sounds when set temperature is reached and elapsed time ends
  • Five-digit LED display can be set for either °C or °F
  • Choose from a wide range of interchangeable aluminum block inserts
This benchtop block chiller/heater is ideal for incubating sample tubes at temperatures ranging from 0 to 40 °C (32 to 104 °F). The Isotemp® temperature control ensures all contents are kept at an even temperature, whether cold or warm. Integrated countdown/count-up timer with audible alarm allows up to nine days to be programmed and will not start until a stable temperature has been reached.
Use with Dri-Block® aluminum inserts to maintain temperature stability of ±0.2 °C. Unit holds two single-width inserts or one double-width inserts. A dark plastic lid protects light-sensitive reagents as well as helping to maintain the sample temperature.
Ordering information: Block(s) not included and sold separately. Interchangeable blocks are available (order separately). Contact your Avantor representative for more information.
Delivery information: Supplied with lid and power cords with plug (US, UK, and EU).