Sodium hydroxide 6.25 Normal
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
R7467000-20F
R7467000-4A
RCR7467000-4AEA
168.51
USD
RCR7467000-4A
RCR7467000-20F
Sodium hydroxide 6.25 Normal
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol Melting Pt: ∼ 0°C Density: 1.23 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 6.245-6.255 N at 20°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...