SHuffle T7 Express lysY Competent E. coli, New England Biolabs
Supplier: New England Biolabs
C3030J
103391-808EA 264 USD
103391-808
SHuffle T7 Express lysY Competent E. coli, New England Biolabs
Competent Cells
Chemically competent E. coli cells suitable for T7 protein expression with enhanced capacity to correctly fold proteins with multiple disulfide bonds in the cytoplasm and with enhanced reduction of basal expression.
  • T7 expression
  • Engineered E. coli B strain to promote disulfide bond formation in the cytoplasm
  • Control of T7 RNA Polymerase by lysozyme allows potentially toxic genes to be expressed
  • Deficient in proteases Lon and OmpT
  • Resistant to phage T1 (fhuA2)