Acetone ≥99.5%, Epredia™ for histology Dehydrant

Supplier: Epredia
Danger

Synonyms: 2-PropanoneDMKPropanoneDimethyl ketone

9011 9011
72060-060CS 394.48 USD
72060-060 CA72060-060
Acetone ≥99.5%, Epredia™ for histology Dehydrant
Acetone

Packaged in easy-to-pour high-density polyethylene containers, Epredia™ High-Quality Dehydrants are safer and lighter than glass bottles.


Formulated to rigid specifications, providing laboratories with high-quality dehydrants.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Flash Pt: <–20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR