Pro-Surface® Disinfectant Wipes with TotalClean™ Technology, Medicom
Supplier: Medicom
40061 40061C 40060C 40060
76449-806CS 385.04 USD
76449-806 CA76449-806 76449-804 CA76449-804
Pro-Surface® Disinfectant Wipes with TotalClean™ Technology, Medicom
Wipes Disposable Wipes and Towels Disinfectant Wipes
Kills the coronavirus in 60 seconds.
  • 1 minute surface contact time accelerates workflow and reduces patient transition time
  • Broad spectrum bactericidal activity eliminates 48 pathogens, including bacteria, viruses, and fungi
  • Low toxicity profile will not damage non-porous instruments or surfaces; no special handling/protective eyewear required
  • Fragrance-free formulation increases compliance without disrupting office operations
  • Low quaternary ammonium content does not leave residue on surfaces
  • Durable, extra-strong wipes cover a large surface area, minimizing consumption; leaves no lint residue
Used to clean or disinfect hard, non-porous surfaces in dental offices, hospitals, and other healthcare settings.
Proprietary TotalClean™ technology provides 1-step cleaning and disinfection in a best-in-class contact time of just 1 minute. Eliminates 48 pathogens, including bacteria, fungi, TB, and viruses, including the coronavirus, without damaging surfaces. Treated surface must remain visibly wet for one minute.
Active ingredient: Ethyl alcohol 72.50% w/w, Didecyl dimethyl ammonium chloride 0.33% w/w, other ingredients 27.17% w/w.