Boron carbide, powder 8 µm
Supplier: Electron Microscopy Sciences
|
Warning
|
50510-08
101411-936EA
112.23
USD
101411-936
Boron carbide, powder 8 µm
Boron carbide
Boron Carbide is one of the hardest materials known, ranking third behind diamond and cubic boron nitride.
- Extreme hardness, (Knoop 100g: 2900-3580 kg/mm2)
- Good chemical resistance
- Good nuclear properties
- Low density (2.52 g/cm2)
Due to the hardness, boron carbide powder is used as an abrasive in polishing and lapping applications.
Formula:
CB₄ |
MDL Number:
MFCD00011520 CAS Number: 12069-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...