Polyethylene glycol 1,000 waxy
Supplier: Electron Microscopy Sciences
Synonyms:
PEG 1,000
, PEG 900-1100
19740
100504-212EA
114.12
USD
100504-212
Polyethylene glycol 1,000 waxy
Polyethylene glycol 1,000
An embedding medium for microscopy and histochemistry
Formula:
H(OCH₂CH₂)nOH MW: 1000 g/mol Density: 1.0926 (60 °C) |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...