Polyethylene glycol 300 viscous liquid

Supplier: Electron Microscopy Sciences

Synonyms: PEG 300

19710
100504-206EA 54.21 USD
100504-206
Polyethylene glycol 300 viscous liquid
Polyethylene glycol 300

An embedding medium for microscopy and histochemistry

Formula: H(OCH₂CH₂)nOH
MW: 300 g/mol
Density: 1.1250 (20 °C)
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR