GelRed® Agarose LE, Biotium
Supplier: Biotium
41029-50G 41029-5G
76403-986EA 326.66 USD
76403-986 76414-950
GelRed® Agarose LE, Biotium
Electrophoresis Gels Agarose and Acrylamide
Ultrapure molecular biology grade agarose pre-coated with GelRed® Nucleic Acid Gel Stain for convenient and safer preparation of precast gels.
  • Non-toxic GelRed® DNA/RNA dye is already in the agarose
  • No need to handle concentrated dye solutions, for enhanced safety
  • Use with TAE or TBE for 0.8 to 2% gels
  • Low EEO, ultrapure molecular biology grade agarose
GelRed® Agarose LE is our ultra-pure molecular biology grade LE agarose pre-coated with GelRed® Nucleic Acid Gel Stain. There is no need to handle concentrated fluorescent dye while preparing your gel,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health
Environmental Certification Classified as non-hazardous to aquatic life under CCR title 22 regulations