DMEM with Glutamine XL, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
112-300-101 112-300-101CS
76470-182EA 31.57 USD
76470-182 76470-184
DMEM with Glutamine XL, Quality Biological
Cell Culture Media
DMEM (Dulbecco's Modified Eagle Medium) Basal Medium with stable L-Alanyl-glutamine (Glutamine XL).
  • Stabilized solution that prevents degradation and cytoxicity
  • Long shelf-life
  • Sterile filtered
DMEM is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells. Quality Biological offers a variety of DMEM modifications for a range of cell culture applications.
Standard, unstabilized solutions of L-glutamine spontaneously break down over time to release cytotoxic amines. Glutamine XL is a stabilized solution that prevents degradation and cytotoxicity, with the benefit of a long shelf life.
The incorporated L-alanine protects the unstable alpha amino acid group of L-glutamine. Aminopeptidase within the cells cleaves the dipeptide, gradually releasing both L-glutamine and L- alanine for use.