CleanDTR Dye Terminator Removal Beads, Bulldog Bio
Supplier: Bulldog Bio
CDTR005 CDTR050 CDTR500
103881-426EA 273.46 USD
103881-426 103881-428 103881-430
CleanDTR Dye Terminator Removal Beads, Bulldog Bio
Nucleic Acid Reagents Sanger Sequencing Reagents
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis.
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis. The process involves just three simple steps: bind, wash, and elute.
Works with BigDye versions 1, 1.1, 2, 3, 3.1 and DYEnamic ET.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more