CellCover Biomolecule Stabilizing Solutions, Bulldog Bio
Supplier: Bulldog Bio
800-050 800-125 800-250 900-196
103881-396EA 200 USD
103881-396 103881-398 103881-400 103881-402
CellCover Biomolecule Stabilizing Solutions, Bulldog Bio
Nucleic Acid Storage Systems
CellCover maintains a cell’s native state offering instant 'freezing' of proteins, RNA, and DNA in their cellular context. It is ideal for preserving morphology, mRNA expression patterns and protein epitopes for furhter analysis, among other uses.
  • Maintain cellular shape integrity without chemical crosslinking
  • Protect DNA, RNA, and proteins
  • Instant, 1-step protocol
  • Non-toxic and ready to use solution
  • Compatible with staining
  • Works with cells in suspension or adherence
  • Works with tissue, tumors, spheroids, and organoids
For parallel storage of proteins,More Product Information