Buchner Funnels, Table Top, PE, Porous, 18", Chemglass
Supplier: Chemglass
CG-1413-05 CG-1413-06 CG-1413-07 CG-1413-08
16002-714EA 1999.33 USD
16002-714 16002-716 16002-718 16002-720
Buchner Funnels, Table Top, PE, Porous, 18", Chemglass
Funnels Filtration Funnels Büchner Funnels
Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate.
  • Removable hose barb connector accepts 12.7 mm (¹/₂") Int.Ø tube and threads into 3/4" NPT bung on the funnel
  • Fixed or removable filter plates are supported by a multiple ring grid
  • HDPE, 6.4 mm (¹/₄") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used with porous filter plates
  • Use temperature range maximum 52 °C (125 °F), minimum –20 °C (–4 °F)
Available in two porosities (medium and coarse); medium isMore Product Information