Diversey™ Oxivir® 1 Wipes, Characteristic Scent, Essendant
Supplier: Janitorial Supplies
DVO100850922 DVO100850923 DVO100850924 DVO100850925 DVO100962573
76461-968CT 233.29 USD
76461-968 76461-970 76461-972 76461-974 76461-984
Diversey™ Oxivir® 1 Wipes, Characteristic Scent, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Next generation ready-to-use disinfectant cleaning wipes are based on proprietary hydrogen peroxide (AHP®) technology to deliver fast, effective cleaning performance. Disinfects in one minute or less.
  • Fast acting, convenient formula, suitable for daily use, and contact time of one minute or less
  • Kills MRSA and Norovirus
  • Formulated with patented AHP® technology containing no phosphates, quats, or phenols
  • Specially designed to penetrate the toughest soils
  • Harnesses the cleaning power of hydrogen peroxide
  • Single-ply wipes
Short contact times of 30 second virucidal (one minute for canine parvovirus); one minute bactericidal, tuberculocidal, and fungicidal; 10 second non-food contact sanitizing and 30 second soft surface sanitizing.
Disinfecting wipes can be used in animal treatment facilities, hotels, long-term care facilities, medical facilities, operating rooms, public buildings, public restrooms, recreational facilities, retail businesses, schools, and vessels; Chemical Compound: Hydrogen Peroxide; Number of Plies: 1.