VWR® Microporous Material Coveralls – Made in USA
Supplier: VWR International
76457-596 76457-598 76457-600 76457-602 76457-604 76457-606 76457-608 76457-610 76457-612 76457-614 76457-616 76457-618 76457-620 76457-622 76457-624 76457-626 76457-628 76457-630 76457-632 76457-634 76457-636 76457-638 76457-640 76457-642 76457-644 76457-646 76457-648 76457-650 76457-652 76457-654 76470-630 76470-632 76470-634 76470-636 76470-638 76470-640 76470-642 76470-644 76470-646 76470-648 76470-650 76470-652 76470-654 76470-656 76470-658 76470-660 76470-662 76470-664 76470-666 76470-668
76457-596CS 514.66 USD
76457-596 76457-598 76457-600 76457-602 76457-604 76457-606 76457-608 76457-610 76457-612 76457-614 76457-616 76457-618 76457-620 76457-622 76457-624 76457-626 76457-628 76457-630 76457-632 76457-634 76457-636 76457-638 76457-640 76457-642 76457-644 76457-646 76457-648 76457-650 76457-652 76457-654 76470-630 76470-632 76470-634 76470-636 76470-638 76470-640 76470-642 76470-644 76470-646 76470-648 76470-650 76470-652 76470-654 76470-656 76470-658 76470-660 76470-662 76470-664 76470-666 76470-668
VWR® Microporous Material Coveralls – Made in USA
Disposable Garments
The VWR® Made in USA PPE line has been expanded to include microporous material coveralls.
  • Microporous, 60 gsm, two-layer material
  • Fluid and particulate barrier
  • Excellent breathability
  • Serged seams
  • Hood and Boot options
  • Elastic or open cuff and ankle options
Microporous material coveralls provide increased fluid protection with great comfort. Choose from a variety of styles, including different wrist and ankle options, with or without hoods, boots, or both.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats,More Product Information