Anti-S protein RBD HEK293 Monoclonal Antibody IgM

Supplier: ACROBIOSYSTEMS
SPDM1411MG SPDM141100UG
103870-908EA 2692.31 USD
103870-908 103870-910
Anti-S protein RBD HEK293 Monoclonal Antibody IgM
Antibodies
Type: Primary
Antigen: S protein RBD
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgM | Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR