Anti-Nucleocapsid Protein Hybridoma Polyclonal Antibody (Trehalose free) (AS60)

Supplier: ACROBIOSYSTEMS
NUNS601MG NUNS60100UG
103870-838EA 2692.31 USD
103870-838 103870-840
Anti-Nucleocapsid Protein Hybridoma Polyclonal Antibody (Trehalose free) (AS60)
Antibodies
Type: Primary
Antigen: Nucleocapsid Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: Mouse IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR