Anti-DNP HEK293 Polyclonal Antibody IgG4

Supplier: Acro Biosystems
DNPM3100UG DNPM31MG
103815-532EA 172.31 USD
103815-532 103815-534
Anti-DNP HEK293 Polyclonal Antibody IgG4
Antibodies
Type: Primary
Antigen: DNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgG4/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR