Sign, 'NOTICE USE HAND SANITIZER BEFORE ENTERING' Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
MRST830VS MRST830VA MRST830VP MRST832VS MRST832VA MRST832VP
76442-498EA 10.68 USD
76442-498 76442-500 76442-502 76442-504 76442-506 76442-508
Sign, 'NOTICE USE HAND SANITIZER BEFORE ENTERING' Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.