Sign, 'DID YOU WASH THEM? HAND WASHING STOPS THE SPREAD…' Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
MRST541VS MRST541VA MRST541VP
76442-378EA 16.05 USD
76442-378 76442-380 76442-382
Sign, 'DID YOU WASH THEM? HAND WASHING STOPS THE SPREAD…' Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.