Preventive Sign, 'PLEASE WEAR FACE MASK THANK YOU' (Green Border) Accuform®
Supplier: Accuform
MPPA510VP MPPA512VA MPPA512VP MPPA510VA MPPA512VS MPPA510VS
76442-310EA 15.42 USD
76442-310 76442-312 76442-304 76442-314 76442-306 76442-308
Preventive Sign, 'PLEASE WEAR FACE MASK THANK YOU' (Green Border) Accuform®
Signs
Signs inform those to keep healthy, safe, and how to help prevent the spread of COVID-19, the Coronavirus.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.