Social Distance Sign, 'Social Distance Maintain 6 ft.' (People with Arrows Graphic) Accuform®
Supplier: Accuform
MGNF544VA MGNF544VS MGNF546VA MGNF544VP MGNF546VS MGNF546VP
76442-058EA 17.15 USD
76442-058 76442-060 76442-062 76442-054 76442-064 76442-056
Social Distance Sign, 'Social Distance Maintain 6 ft.' (People with Arrows Graphic) Accuform®
Signs
Signs alert and remind those to practice social and physical distancing to prevent the spread of COVID-19, the Coronavirus.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.