Sign, 'WARNING BIOSAFETY LEVEL 2' Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
MBHZ322VS MBHZ322VA MBHZ322VP
76441-816EA 17.68 USD
76441-816 76441-818 76441-820
Sign, 'WARNING BIOSAFETY LEVEL 2' Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.