Sign, 'DANGER WAREHOUSE CLOSED, DISINFECTANT AEROSOL IN PROGRESS' Accuform®
Supplier: Accuform
MBHZ135VA MBHZ137VP MBHZ135VP MBHZ137VA MBHZ137VS MBHZ135VS
76441-776EA 28.75 USD
76441-776 76441-774 76441-784 76441-778 76441-782 76441-780
Sign, 'DANGER WAREHOUSE CLOSED, DISINFECTANT AEROSOL IN PROGRESS' Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.