Medical Power Strips, Tripp Lite
Supplier: Tripp Lite
PS-615-HG PS-415-HGULTRA PS-606-HG PS-402-HG-OEM PS-410-HGOEMCC PS-415-HG PS-415-HG-OEM PS-615-HG-OEM SPS415HGULTRA PS625HG20AOEM PS615HG20AOEM PS607HG20AOEM PS-607-HG-OEM PS-407-HG-OEM PS-406-HGULTRA SPS406HGULTRA PS-602-HG
89079-128EA 192.96 USD
89079-128 89079-130 89079-132 10144-226 10144-228 10144-232 10144-234 10144-238 10144-444 76461-410 76461-412 76461-414 76461-416 76461-418 76461-420 76461-422 76461-424 76461-426 76461-428
Medical Power Strips, Tripp Lite
Electrical Sockets
Medical-grade power strips designed and manufactured in accordance with Article 517 and certified to meet UL standards.
  • Power distribution anywhere in any medical facility
  • Exclusive, patented antimicrobial protection
  • Versatile medical-grade solution
  • Premium safety features
  • Ready to use right out of the box
The Medical-Grade Power Strip supplies power to movable plug-connected components mounted to a rack, table or pedestal in operating rooms, patient-care vicinities, therapy rooms, imaging environments and administrative areas. Compliant with UL 1363A specifications, the power strip features a NEMA 5-15P-HG hospital-grade plug and NEMA 5-15R-HG hospital-grade outlets that include integrated locking safety covers.
The power strip is protected by a patented antimicrobial powder coating with EPA-registered and FDA-compliant silver ionic technology. The antimicrobial surface protection is 99.9 percent effective in inhibiting the growth of bacteria, such as C. diff and MRSA, and reducing the risk of HAIs in hospitals and other medical environments. The coating meets the requirements of JIS Z 2801:2000, an international standard for evaluating efficacy in antimicrobial products.
Built-in resettable circuit breakers control power to all outlets and protect the power strip against overloads. Switchless design prevents the unit from being accidentally shut off. The power cord provides ample reach to the AC power source.
Mounting tabs and flanges let you install the power strip on a wide variety of surfaces, such as IV poles, mobile carts, crash carts and mobile medical workstations. The all-metal housing ensures safety and is guaranteed to last a long time in demanding environments.
Certifications: UL 1363A.