Edvotek What is an Epidemic and How Does Infection Spread?
Supplier: Edvotek
S-68
470331-024EA 99.8 USD
470331-024
Edvotek What is an Epidemic and How Does Infection Spread?
Educational Materials Biology Educational Materials Microbiology Learning Activities
Explore how bacteria and viruses spread through a population.
  • NGSS-aligned with MS-LS2
  • Use clear solutions and a color-changing indicator to model a classroom infection
  • Record potential infection events and practice contract tracing
  • Discuss different types of infections and different detection techniques
  • Contains everything you need to complete the activity in 30 min!
Infectious agents such as bacteria and viruses can spread rapidly through a population and cause widespread disease and death. In this experiment, your students will use colored solutions to simulate the spreading of a disease in the classroom.
Ordering information: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Caution: Components may be stored at room temperature.