BHD Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BWLNWB24 BWLNWB30 BWLNWB36 BWLNWB48
76450-320CS 3093.75 USD
76450-320 76450-322 76450-324 76450-328
BHD Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Individually poly bagged
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable