Epredia™ Solvent-Resistant White Biopsy Bags, Epredia
Supplier: Epredia
6774022 6774024 6774019
10170-228CS 915.49 USD
10170-228 10170-230 10171-088
Epredia™ Solvent-Resistant White Biopsy Bags, Epredia
Bags Sample Bags
Made of thin-mesh solvent resistant nylon providing for optimal fluid exchange around samples.
  • Minimize the risk of small specimen loss
  • Rapidly remove specimens with convenient peel-open bag
Designed to keep tissues safe while allowing for the even exchange of fluids. Available in a range of sizes to meet specific needs.